missing translation for 'onlineSavingsMsg'
Learn More

Tozadenant, MedChemExpress

Product Code. 16454338
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454338 10 mg 10mg
16427057 5 mg 5mg
16425256 100 mg 100mg
16410667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16454338 Supplier Medchem Express Supplier No. HY10995
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tozadenant is an adenosine A2A receptor antagonist, with Ki of 11.5 nM on human A2A and 6 nM on rhesus A2A.

Tozadenant is an adenosine A2A receptor antagonist, with Ki of 11.5 nM on human A2A and 6 nM on rhesus A2A.

Specifications

Chemical Name or Material Tozadenant
CAS 870070-55-6
Color White
Purity Grade Notes Research
Molecular Formula C19H26N4O4S
Quantity 10 mg
Synonym SYN115
Solubility Information DMSO : 50 mg/mL (123.00 mM; Need ultrasonic)
SMILES O=C(NC1=NC2=C(C=CC(N3CCOCC3)=C2S1)OC)N4CCC(O)(CC4)C
Molecular Weight (g/mol) 406.5
Formula Weight 406.5
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.