missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPCN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94265-0.1ml
This item is not returnable.
View return policy
Description
TPCN1 Polyclonal antibody specifically detects TPCN1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| TPCN1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ20612, KIAA1169, TPC1, two pore segment channel 1, two-pore channel 1, two-pore segment channel 1, voltage-dependent calcium channel protein TPC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 692-816 of human TPCN1 (NP_060371.2). FRMNYSRKNQDSEVDGGITLEKEISKEELVAVLELYREARGASSDVTRLLETLSQMERYQQHSMVFLGRRSRTKSDLSLKMYQEEIQEWYEEHAREQEQQRQLSSSAAPAAQQPPGSRQRSQTVT | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 53373 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction