missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPMT Antibody (SR1518), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 449.00 €
Specifications
| Antigen | TPMT |
|---|---|
| Clone | SR1518 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630844
|
Novus Biologicals
NBP3-22563-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627094
|
Novus Biologicals
NBP3-22563-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPMT Monoclonal antibody specifically detects TPMT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| TPMT | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
| A synthesized peptide derived from human TPMT (Uniprot #: P51580) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1518 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Proteases & Other Enzymes, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7172 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title