missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPO (Thyroid Peroxidase) (Thyroid Marker) Mouse, Clone: TPO/1922, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7173-MSM2-P1
This item is not returnable.
View return policy
Specifications
| TPO (Thyroid Peroxidase) (Thyroid Marker) | |
| Cytoplasmic and Cell Surface | |
| TPO/1922 | |
| Unconjugated | |
| Mouse | |
| 100kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 7173 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human TPO (apprx. aa 685-804) | |
| Thyroid Carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction