missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPP2 Recombinant Rabbit Monoclonal Antibody (24GB2015)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00 € - 564.00 €

Specifications

Antigen TPP2
Clone 24GB2015
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276127
GREENER_CHOICE
View Documents
Invitrogen™
MA557798
100 μL
564.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275630
View Documents
Invitrogen™
MA557799
20 μL
198.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TPP2 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
Specifications

Specifications

TPP2
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
TPP2
TPP2; TPP-2; TppII; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase 2; tripeptidyl peptidase II; tripeptidyl-peptidase 2; Tripeptidyl-peptidase II
TPP2
Primary
Affinity chromatography
24GB2015
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P29144, Q64514, Q64560
22019, 7174, 81815
A synthesized peptide derived from human Tripeptidyl peptidase II (aa 900-1150).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.