missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPX2 Recombinant Rabbit Monoclonal Antibody (K01_2P81)

Product Code. 30830341
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830341 50 μL
30824765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830341 Supplier Invitrogen™ Supplier No. MA565682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TPX2 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPX2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2P81
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Tpx2
Gene Accession No. Q9ULW0
Gene Alias 2610005B21Rik; C20orf1; C20orf2; Differentially expressed in cancerous and non-cancerous lung cells 2; differentially expressed in lung cells; DIL2; DIL-2; FLS353; GD: HCA519; GD:C20orf1; HCA519; HCTP4; Hepatocellular carcinoma-associated antigen 519; hepatocellular carcinoma-associated antigen 90; p100; preferentially expressed in colorectal cancer; protein fls353; REPP86; restricted expression proliferation associated protein 100; restricted expression proliferation-associated protein 100; Targeting protein for Xklp2; TPX 2; Tpx2; TPX2 microtubule nucleation factor; TPX2, microtubule nucleation factor; TPX2, microtubule-associated; TPX2, microtubule-associated protein homolog; TPX2, microtubule-associated protein homolog (Xenopus laevis); TPX2, microtubule-associated, homolog
Gene Symbols Tpx2
Host Species Rabbit
Immunogen Recombinant protein of human TPX2.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22974
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.