missing translation for 'onlineSavingsMsg'
Learn More

TR beta 1/NR1A2/Thyroid Hormone Receptor beta Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00 € - 470.00 €

Specifications

Antigen TR beta 1/NR1A2/Thyroid Hormone Receptor beta
Dilution Western Blot 1:500 - 1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18629831
View Documents
Novus Biologicals
NBP2-93985-0.02ml
0.02 mL
190.00 €
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673032
View Documents
Novus Biologicals
NBP2-93985-0.1ml
0.1 mL
470.00 €
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TR beta 1/NR1A2/Thyroid Hormone Receptor beta Polyclonal antibody specifically detects TR beta 1/NR1A2/Thyroid Hormone Receptor beta in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

TR beta 1/NR1A2/Thyroid Hormone Receptor beta
Western Blot
Unconjugated
Rabbit
Cancer, Neuroscience
PBS (pH 7.3), 50% glycerol
7068
IgG
Affinity purified
Western Blot 1:500 - 1:1000
Polyclonal
Purified
RUO
Human, Mouse, Rat
avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, c-erbA-2, c-erbA-beta, ERBA2MGC126109, ERBA-BETA, GRTH, MGC126110, NR1A2thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog 2), Nuclear receptor subfamily 1 group A member 2, oncogene ERBA2, PRTH, THR1pituitary resistance to thyroid hormone, THRB1, THRB2, thyroid hormone receptor beta, thyroid hormone receptor beta 1, thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a)oncogene homolog 2, avian)
Recombinant fusion protein containing a sequence corresponding to amino acids 1-263 of human TR beta 1/NR1A2/Thyroid Hormone Receptor beta (NP_000452.2). MTPNSMTENGLTAWDKPKHCPDREHDWKLVGMSEACLHRKSHSERRSTLKNEQSSPHLIQTTWTSSIFHLDHDDVNDQSVSSAQTFQTEEKKCKGYIPSYLDKDELCVVCGDKATGYHYRCITCEGCKGFFRRTIQKNLHPSYSCKYEGKCVIDKVTRNQCQECRFKKCIYVGMATDLVLDDSKRLAKRKLIEENREKRRREELQKSIGHKPEPTDEEWELIKTVTEAHVATNAQGSHWKQKRKFLPEDIGQAPIVNAPEGGK
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.