missing translation for 'onlineSavingsMsg'
Learn More

TRA-1-60 (Podocalyxin) Mouse, anti-Human, PE, Clone: TRA-1-60R, STEMCELL Technologies

Mouse Monoclonal Antibody

274.05 € - 450.90 €

Specifications

Antigen TRA-1-60 (Podocalyxin)
Clone TRA-1-60R
Applications Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17168801
Encompass_Preferred
View Documents
Stemcell Technologies
60064PE.1
25 Tests
274.05 €
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17158801
Encompass_Preferred
View Documents
Stemcell Technologies
60064PE
100 Tests
450.90 €
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The TRA‐1‐60R antibody reacts with TRA-1-60 (Podocalyxin), a > 200 kDa pluripotent stem cell-specific protein expressed on the surface of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells, embryonal carcinoma (EC) cells and embryonic germ (EG) cells, as well as rhesus monkey ES cell lines
  • The epitope contains sialic acid and is associated with a large-molecular-mass transmembrane protein named podocalyxin
  • Though sialylated, the epitope recognized by the TRA‐1‐60R antibody is resistant to treatment with neuraminidase
  • This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860)
Specifications

Specifications

TRA-1-60 (Podocalyxin)
Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
PE
Rhesus Monkey, Human, Rabbit
Primary
TRA-1-60R
Monoclonal
Mouse
IgM κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.