missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Trabikibart Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

477.00 € - 1204.00 €

Specifications

Antigen Trabikibart
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284552
GREENER_CHOICE
View Documents
Invitrogen™
MA558863
100 μg
477.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283296
View Documents
Invitrogen™
MA558864
1 mg
1204.00 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Trabikibart, also known as CSL311, is a fully human monoclonal antibody that targets the common beta chain (beta c) receptor, shared by the interleukin-3 (IL-3), granulocyte-macrophage colony-stimulating factor (GM-CSF), and interleukin-5 (IL-5) cytokines. Colony stimulating factor 2 receptor subunit beta (CSF2RB), also known as CD131, is a critical component of the cytokine receptor family involved in hematopoiesis and immune response regulation. Located on chromosome 22q13.1, the CSF2RB gene encodes the common beta-subunit that pairs with the alpha-subunits of receptors for granulocyte-macrophage colony-stimulating factor (GM-CSF), interleukin-3 (IL-3), and interleukin-5 (IL-5). Structurally, the beta-subunit contributes to the formation of high-affinity receptor complexes essential for signal transduction. Functionally, upon ligand binding, CSF2RB stimulates various intracellular signaling cascades that mediate cellular proliferation, differentiation, and survival, particularly of hematopoietic progenitor cells and immune cells. Aberrations in CSF2RB expression or function can lead to immune dysfunctions and are associated with diseases such as asthma and leukemia. Recent studies have highlighted the importance of the CSF2RB in regulatory T cells, underscoring its role in autoimmune disease prevention.
Specifications

Specifications

Trabikibart
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG4SP
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.