missing translation for 'onlineSavingsMsg'
Learn More

Trace Metals 1 For Environmental Analysis, SPEX CertiPrep™

Product Code. 15698335 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15698335 500 mL 500mL
15608345 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15698335 Supplier SPEX Certiprep Supplier No. WP15500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al500μg/mL
V250μg/mL
As100μg/mL
Be100μg/mL
Co100μg/mL
Cr100μg/mL
Cu100μg/mL
Fe100μg/mL
Mn100μg/mL
Ni100μg/mL
Pb100μg/mL
Zn100μg/mL
Cd25μg/mL
Se25μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Trace Metals I
Name Note Groundwater and Wastewater Pollution Control Check Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 5% HNO3
Industry Type Wastewater Testing
Quantity 500 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Various
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.