missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™

Product Code. 11754246 Shop All Fisherbrand Products
Change view
Click to view available options
Conductivity:
10, 100, 1000, 1413, 10,000, 100,000 μ℧
10,000 μ℧
100 μ℧
100,000 μ℧
1000 μ℧
1413 μ℧
Quantity:
100 mL
6 x 100 mL
Unit Size:
Pack of 6
Product Code. Conductivity Quantity unitSize
11754246 1413 μ℧ 6 x 100 mL Pack of 6
11774246 100 μ℧ 6 x 100 mL Pack of 6
11784246 1000 μ℧ 100 mL Pack of 6
11794246 10,000 μ℧ 100 mL Pack of 6
11704256 100,000 μ℧ 100 mL Pack of 6
11744246 10, 100, 1000, 1413, 10,000, 100,000 μ℧ 6 x 100 mL Pack of 6
6 options

Product Code. 11754246

Please to purchase this item. Need a web account? Register with us today!

Ideal for lab or field applications

Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.

Calibration within container — zero contamination

  • Traceable to NIST (National Institute of Standards and Technology)
  • Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
  • 100% compatible with all conductivity meters
  • A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
  • Includes an individual temperature chart and step-by-step calibration instructions
  • Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
  • Single use Eliminates external contamination concerns Each standard contains 100mL

Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12

Order Info

Each bottle contains 100mL

Specifications

Chemical Name or Material Conductivity Standard
Packaging 6/Pk.
Conductivity 1413 μ℧
Technique Calibration of Conductivity Instrument
Color Clear
pH 7
For Use With (Equipment) Conductivity Meter
Solubility Soluble in water
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 6 x 100 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.