missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TraceGOLD™ TG-WaxMS A GC Columns

Product Code. 10417724 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
15 m
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
1.5 μm
Unit Size:
Each
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
10417724 30 m 0.53 mm 1 μm Each
10086254 15 m 0.25 mm 0.25 μm Each
10594475 15 m 0.32 mm 0.25 μm Each
10754057 30 m 0.25 mm 0.25 μm Each
10229274 30 m 0.32 mm 0.25 μm Each
10147604 30 m 0.53 mm 0.25 μm Each
10535615 60 m 0.25 mm 0.25 μm Each
10660426 60 m 0.53 mm 0.25 μm Each
10596565 15 m 0.25 mm 0.5 μm Each
10468464 15 m 0.32 mm 0.5 μm Each
10596755 15 m 0.53 mm 0.5 μm Each
10487314 30 m 0.25 mm 0.5 μm Each
10781937 30 m 0.32 mm 0.5 μm Each
10791937 30 m 0.53 mm 0.5 μm Each
10487504 60 m 0.25 mm 0.5 μm Each
10628515 60 m 0.32 mm 0.5 μm Each
10499424 60 m 0.53 mm 0.5 μm Each
10754837 15 m 0.32 mm 1 μm Each
10640256 15 m 0.53 mm 1 μm Each
10220245 30 m 0.32 mm 1 μm Each
10427724 60 m 0.32 mm 1 μm Each
10095934 60 m 0.53 mm 1 μm Each
10409434 15 m 0.53 mm 1.5 μm Each
10229554 30 m 0.53 mm 1.5 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Product Code. 10417724 Supplier Thermo Scientific™ Supplier No. 260872980
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage with this low-bleed, polar GC column.

Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage using Thermo ScientificTM TraceGOLDTM TG-WaxMS A GC column.

Engineered for Superior Performance
  • Polar phase, acid-deactivated polyethylene glycol
  • Resists oxidative damage and adsorption of acids
  • Excellent peak shapes for high MW acids
  • Equivalent to USP G25, G35 phases
Applications:
  • Organic acids
  • Free fatty acids
  • Alcohols

A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.

Recommended For:
  • Acids
  • Carboxylic acids
  • Organic acids
  • Alternative Uses:
  • Alcohols
  • Alcohols in beverages
  • Aldehydes
  • Alditol acetates (sugars)
  • Essential oils
  • Similar To:
  • DB-FFAP
  • HP-FFAP
  • Nukol™
  • OV™-351
  • CP-Wax 58 CB
  • Stabilwax™-DA
  • BP-21
  • Optima™ FFAP
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Alcohol Analysis, GC/MS for Free Fatty Acids, GC/MS for Alcohols
Film Thickness 1 μm
Length (Metric) 30 m
Polarity Polar
Description 1 um Thickness; 0.53 mm ID; 30 m Length
Diameter (Metric) 0.53 mm
USP Type G25, G35
Stationary Phase TraceGOLD™ TG-WAXMS A
Diameter (Metric) Inner 0.53 mm
Product Line TraceGOLD
Max. Temperature (Metric) 240/250°C
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.