missing translation for 'onlineSavingsMsg'
Learn More

Tradipitant, MedChemExpress

Product Code. 16439697
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439697 5 mg 5mg
16437168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16439697 Supplier Medchem Express Supplier No. HY16732
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tradipitant (VLY-686) is a neurokinin-1 (NK-1) antagonist.

Tradipitant (VLY-686) is a neurokinin-1 (NK-1) antagonist.

Specifications

Chemical Name or Material Tradipitant
Physical Form Solid
CAS 622370-35-8
Color Tan
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C28H16ClF6N5O
For Use With (Application) Neuroscience-Neuromodulation
Synonym VLY-686 LY686017
Solubility Information DMSO : 100 mg/mL (170.10 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1C2=C(C3=CC=NC=C3)N(CC4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)N=N2)C5=CC=CC=C5Cl
Molecular Weight (g/mol) 587.9
Formula Weight 587.9
Percent Purity 99.34%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.