missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRAILR4/TNFRSF10D/DcR2 Antibody Pair [HRP]

Product Code. p-7183348 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18250077

Brand: Novus Biologicals™ NBP27945515PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRAILR4/TNFRSF10D/DcR2.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 234.38 pg/mL
Assay Sensitivity 234.38-15000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRAILR4/TNFRSF10D/DcR2.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain
Gene ID (Entrez) 8793
Gene Symbol TNFRSF10D
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human TRAILR4/TNFRSF10D/DcR2 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.3 mg/mL of mouse anti-Human TRAILR4/TNFRSF10D/DcR2 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 345 ng of recombinant Human TRAILR4/TNFRSF10D/DcR2.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target TRAILR4/TNFRSF10D/DcR2
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 15000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.