missing translation for 'onlineSavingsMsg'
Learn More

TRAM-34, MedChemExpress

Product Code. 16441887
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441887 50 mg 50mg
16415728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441887 Supplier Medchem Express Supplier No. HY13519
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TRAM-34 is a highly selective blocker of intermediate-conductance calcium-activated K+ channel (IKCa1) (Kd=20 nM).

TRAM-34 is a highly selective blocker of intermediate-conductance calcium-activated K+ channel (IKCa1) (Kd=20 nM).

Specifications

Chemical Name or Material TRAM-34
Physical Form Powder
CAS 289905-88-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H17ClN2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 25 mg/mL (72.50 mM; Need ultrasonic)
SMILES ClC1=CC=CC=C1C(N2N=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
Molecular Weight (g/mol) 344.84
Formula Weight 344.84
Percent Purity 99.32%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.