missing translation for 'onlineSavingsMsg'
Learn More

Trametinib, MedChemExpress

Product Code. 16494338
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494338 10 mg 10mg
16450667 50 mg 50mg
16465256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494338 Supplier Medchem Express Supplier No. HY10999
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Trametinib (GSK1120212; JTP-74057) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib activates autophagy and induces apoptosis.

Trametinib (GSK1120212

Specifications

Chemical Name or Material Trametinib
CAS 871700-17-3
Color White
Purity Grade Notes Research
Molecular Formula C26H23FIN5O4
Quantity 10 mg
Synonym GSK1120212 JTP-74057
Solubility Information DMSO : 25 mg/mL (40.62 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(NC1=CC=CC(N(C2=O)C(C(C(N2C3CC3)=O)=C(N4C)NC5=CC=C(C=C5F)I)=C(C4=O)C)=C1)=O
Molecular Weight (g/mol) 615.39
Formula Weight 615.39
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.