missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (12A668) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 27 publications
230.00 € - 538.00 €
Specifications
| Antigen | TRANCE/TNFSF11/RANK L |
|---|---|
| Clone | 12A668 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, ELISA reported in scientific literature (PMID 15935726), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Immunoassay, Flow (Cell Surface), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18757333
|
Novus Biologicals
NB100-56512SS |
0.025 mg |
230.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758903
|
Novus Biologicals
NB100-56512 |
0.1 mg |
538.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoassay, Flow (Cell Surface).Specifications
| TRANCE/TNFSF11/RANK L | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| O14788 | |
| 8600 | |
| A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE/TNFSF11/RANK L was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 12A668 | |
| Western Blot 0.5-2 ug/ml, ELISA reported in scientific literature (PMID 15935726), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Immunoassay, Flow (Cell Surface), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| TNFSF11 | |
| IgG1 κ | |
| Protein G purified | |
| 35 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title