missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dibenzoylethylene, 97%

Product Code. 10550201 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10550201

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 959-28-4 | C16H12O2 | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-28-4
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00003083
InChI Key WYCXGQSQHAXLPK-VAWYXSNFSA-N
Synonym trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
PubChem CID 735960
IUPAC Name (E)-1,4-diphenylbut-2-ene-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 108.0°C to 112.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COCH=CHCOC6H5
Quantity 5 g
Beilstein 07, IV, 2578
Fieser 01,195
Solubility Information Solubility in water: insoluble
Formula Weight 236.27
Percent Purity 97%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material trans-1, 2-Dibenzoylethylene, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.