missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Cyclohexane diisocyanate, 97%

Product Code. 11302837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302837 5 g 5g
11392827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302837 Supplier Thermo Scientific Alfa Aesar Supplier No. L15991.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 7517-76-2 | C8H10N2O2 | 166.18 g/mol

Mixed aromatic/alicyclic polyimides were prepared by polycondensation reactions of trans-1,4-cyclohexane diisocyanate CHDI with PMDA, BTDA, and 6FDA. The intermediate reactivity observed for CHDI, allows preparation of polyurethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mixed aromatic/alicyclic polyimides were prepared by polycondensation reactions of trans-1,4-cyclohexane diisocyanate CHDI with PMDA, BTDA, and 6FDA. The intermediate reactivity observed for CHDI, allows preparation of polyurethanes.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, alcohols, amines.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7517-76-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00074787
InChI Key CDMDQYCEEKCBGR-UHFFFAOYSA-N
Synonym trans-1,4-diisocyanatocyclohexane, trans-1,4-cyclohexane diisocyanate, trans-1,4-cyclohexanediisocyanate, 1,4-cyclohexane diisocyanate, cyclohexane, 1,4-diisocyanato, cyclohexane, 1,4-diisocyanato-, trans, cyclohexane-1,4-diisocyanate, trans-1,4-cyclohexyleneisocyanate, 1,4-cyclohexyl di-isocyanate, cis-1,4-cyclohexylenediisocyanate
PubChem CID 17367
IUPAC Name 1,4-diisocyanatocyclohexane
SMILES C1CC(CCC1N=C=O)N=C=O

Spezifikation

Melting Point 59°C to 62°C
Boiling Point 145°C to 146°C (32 mmHg)
Odor Irritating
Quantity 5 g
UN Number UN2811
Beilstein 2804862
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material trans-1,4-Cyclohexane diisocyanate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.