missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Methyl-3-phenyl-2-propen-1-ol, 95%

Product Code. 10356464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356464 25 g Glass bottle 25g
10132834 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356464 Supplier Thermo Scientific Acros Supplier No. 427050250
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1504-55-8 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1504-55-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
InChI Key LLNAMUJRIZIXHF-VQHVLOKHSA-N
Synonym 2-methyl-3-phenyl-2-propen-1-ol, beta-methylcinnamyl alcohol, 2-propen-1-ol, 2-methyl-3-phenyl, alpha-methylcinnamyl alcohol, methyl cinnamic alcohol, alpha-methylcinnamic alcohol, alpha methyl cinnamic alcohol, 3-phenyl-2-methyl-propen-2-ol-1, e-2-methyl-3-phenyl-2-propen-1-ol, 2e-2-methyl-3-phenylprop-2-en-1-ol
PubChem CID 6393846
IUPAC Name (E)-2-methyl-3-phenylprop-2-en-1-ol
SMILES CC(=CC1=CC=CC=C1)CO

Specifications

Density 1.0300g/mL
Boiling Point 77°C (0.1 mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=C(CH3)CH2OH
Quantity 25 g
Specific Gravity 1.03
Formula Weight 148.2
Percent Purity 95%
Chemical Name or Material trans-2-Methyl-3-phenyl-2-propen-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Methyl-3-phenyl-2-propen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.