missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octenoic acid, 94%, Thermo Scientific™

Product Code. p-7058262
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331497 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11331497 Supplier Thermo Scientific Alfa Aesar Supplier No. L02058.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-67-6
Molecular Formula C8H13O2
Molecular Weight (g/mol) 141.19
MDL Number MFCD00002706
InChI Key CWMPPVPFLSZGCY-VOTSOKGWSA-M
Synonym trans-2-octenoic acid, 2-octenoic acid, e-2-octenoic acid, e-oct-2-enoic acid, 2-octenoic acid, 2e, 2e-octenoic acid, 2-octenoic acid, e, trans-alpha-octenoic acid, unii-c0057tj59e, oct-2-enoic acid
PubChem CID 5282714
ChEBI CHEBI:86544
IUPAC Name (E)-oct-2-enoic acid
SMILES CCCCC\C=C\C([O-])=O

Specifications

Density 0.942
Boiling Point 101°C to 102°C (5 mmHg)
Flash Point 140°C (284°F)
Odor Characteristic
Refractive Index 1.4588
Quantity 50 g
UN Number UN3265
Beilstein 1721115
Formula Weight 142.2
Percent Purity 94%
Chemical Name or Material trans-2-Octenoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.