missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Fluorocinnamic acid, 98%

Product Code. 11485746
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485746 100 g 100g
11465746 5 g 5g
11475746 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485746 Supplier Thermo Scientific Alfa Aesar Supplier No. B20818.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20595-30-6 | C9H7FO2 | 166.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20595-30-6
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD00004383
InChI Key RTSIUKMGSDOSTI-SNAWJCMRSA-N
Synonym 3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid
PubChem CID 1551219
IUPAC Name (E)-3-(3-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)F)C=CC(=O)O

Specifications

Melting Point 165°C to 168°C
Odor Odorless
Quantity 100 g
Beilstein 1365172
Formula Weight 166.15
Percent Purity 98%
Chemical Name or Material trans-3-Fluorocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.