missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexen-1-ol, 97%

Product Code. 11348495
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348495 5 g 5g
11358495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348495 Supplier Thermo Scientific Alfa Aesar Supplier No. L10541.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-97-2 | C6H12O | 100.161 g/mol

trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-97-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002960
InChI Key UFLHIIWVXFIJGU-ONEGZZNKSA-N
Synonym trans-3-hexen-1-ol, e-hex-3-en-1-ol, e-3-hexen-1-ol, trans-3-hexenol, 3-hexen-1-ol, 3e-hex-3-en-1-ol, 3-hexen-1-ol, e, trans-hex-3-en-1-ol, unii-4e0nfr5b3u, 3e-hexen-1-ol
PubChem CID 5284503
IUPAC Name (E)-hex-3-en-1-ol
SMILES CCC=CCCO

Specifications

Density 0.845
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Pleasant
Refractive Index 1.439
Quantity 5 g
UN Number UN1987
Beilstein 1719713
Merck Index 14,4700
Solubility Information Slightly soluble in water.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material trans-3-Hexen-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.