missing translation for 'onlineSavingsMsg'
Learn More

trans-4-[4-(4-n-Pentylcyclohexyl)phenyl]benzonitrile, 99%, Thermo Scientific™

Product Code. 15402946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15402946 1 g 1g
15412946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402946 Supplier Thermo Scientific Alfa Aesar Supplier No. H56850.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It finds its application as an intermediate of polymer-dispersed liquid crystals (PDLCs) are a relatively new class of materials that hold promise for many applications ranging from switchable windows to projection displays. These materials, which are simply a combined application of polymers and liquid crystals, are the focus of extensive research in the display industry.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 68065-81-6
Molecular Formula C24H29N
Molecular Weight (g/mol) 331.503
MDL Number MFCD01076317
InChI Key QKEBUASRTJNJJS-UHFFFAOYSA-N
Synonym 4'-trans-4-pentylcyclohexyl-1,1'-biphenyl-4-carbonitrile, trans-4'-4-pentylcyclohexyl-4-biphenylcarbonitrile, trans-4-4-pentylcyclohexyl-4'-cyanobiphenyl, 4-trans-4-n-pentylcyclohexyl biphenyl-4'-carbonitrile, 4-4-4-pentylcyclohexyl phenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-trans-4-pentylcyclohexyl, 4-cyano-4'-trans-4-pentylcyclohexyl biphenyl, 4'-trans-4-amylcyclohexyl biphenyl-4-carbonitrile, trans-4'-4-pentylcyclohexyl 1,1'-biphenyl-4-carbonitrile
PubChem CID 106491
IUPAC Name 4-[4-(4-pentylcyclohexyl)phenyl]benzonitrile
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N

Spécification

Melting Point 95°C to 97°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 331.5
Percent Purity 99%
Chemical Name or Material trans-4-[4-(4-n-Pentylcyclohexyl)phenyl]benzonitrile
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.