missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%

Product Code. 11490347
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490347 10 g 10g
11400357 50 g 50g
11410357 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490347 Supplier Thermo Scientific Alfa Aesar Supplier No. B21742.09
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1197-18-8 | C8H15NO2 | 157.213 g/mol

It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-18-8
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00001466
InChI Key GYDJEQRTZSCIOI-UHFFFAOYSA-N
Synonym tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
PubChem CID 5526
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CN)C(=O)O

Specifications

Melting Point >300°C
Beilstein 2207452
Merck Index 14,9569
Solubility Information Soluble in water (167mg/ml),ethanol (<1mg/ml at 25°C). Very slightly soluble in ether,DMSO (<1mg/ml at 25°C),and alcohol.
Quantity 10 g
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material trans-4-(Aminomethyl)cyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.