accessibility menu, dialog, popup

UserName

CAS RN 1197-18-8

IUPAC Name: 4-(aminomethyl)cyclohexane-1-carboxylic acid
Synonyms: tranexamic acid
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: GYDJEQRTZSCIOI-UHFFFAOYSA-N
SMILES: NCC1CCC(CC1)C(O)=O
Molecular Weight (g/mol) 
  • (10)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (10)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)