missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methoxy-beta-nitrostyrene, 98%

Product Code. 11490703
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490703 25 g 25g
11480703 5 g 5g
11400713 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490703 Supplier Thermo Scientific Alfa Aesar Supplier No. A12745.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5576-97-6 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5576-97-6
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00024826
InChI Key JKQUXSHVQGBODD-UHFFFAOYSA-N
Synonym 1-methoxy-4-2-nitrovinyl benzene, 4-methoxy-beta-nitrostyrene, p-2-nitrovinyl anisole, trans-4-methoxy-beta-nitrostyrene, e-1-methoxy-4-2-nitrovinyl benzene, 2'-nitro vinyl anisole, 1-methoxy-4-e-2-nitroethenyl benzene, 1-4-methoxyphenyl-2-nitroethene, 1-4-methoxyphenyl-2-nitroethylene, 4-2-nitrovinyl anisole
PubChem CID 697963
SMILES COC1=CC=C(C=C[N+]([O-])=O)C=C1

Specifications

Melting Point 86°C to 88°C
Quantity 25 g
Beilstein 2047935
IUPAC Name 1-methoxy-4-[(E)-2-nitroethenyl]benzene
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material trans-4-Methoxy-beta-nitrostyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.