missing translation for 'onlineSavingsMsg'
Learn More

trans-Anethole, 99%

Product Code. 10222942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10222942 500 mL Glass bottle 500mL
10149931 5 mL Glass bottle 5mL
10081850 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10222942 Supplier Thermo Scientific Acros Supplier No. 152535000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4180-23-8 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC

Specifications

Melting Point 23.0°C
Color Colorless to Yellow
Density 0.9875g/mL
Boiling Point 234.0°C to 237.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHC6H4OCH3
Refractive Index 1.5590 to 1.5620
Quantity 500 mL
Beilstein 06, 566
Merck Index 15, 636
Specific Gravity 0.9875
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with ether, chloroform, soluble in benzene, ethyl acetate, acetone,, alcohol and carbon disulfide
Formula Weight 148.2
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material trans-Anethole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.