missing translation for 'onlineSavingsMsg'
Learn More

Melford trans-Cinnamic Acid

Product Code. p-200087287
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 30018143

Brand: Melford C3440025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Auxin like growth regulator.

  • CAS Number: 140-10-3
  • Molecular Formula: C9H8O2
  • Molecular Weight: 148.2

Specifications

CAS 140-10-3
Molecular Formula C9H8O2
Quantity 25 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.