missing translation for 'onlineSavingsMsg'
Learn More

trans-p-Coumaric Acid, TRC

Product Code. 17994285
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994285 25 g 25g
17971375 1 g 1g
17914845 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994285 Supplier Toronto Research Chemicals Supplier No. TRCC75539025G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-98-4
Molecular Formula C9 H8 O3
Molecular Weight (g/mol) 164.16
Synonym 2-Propenoic acid, 3-(4-hydroxyphenyl)-, (2E)- (9CI, ACI), (2E)-3-(4-Hydroxyphenyl)-2-propenoic acid (ACI), 2-Propenoic acid, 3-(4-hydroxyphenyl)-, (E)- (ZCI), Cinnamic acid, p-hydroxy-, (E)- (8CI), (2E)-3-(4-Hydroxyphenyl)prop-2-enoic acid, (E)-3-(4-Hydroxyphenyl)-2-propenoic acid, (E)-3-(4-Hydroxyphenyl)acrylic acid, (E)-3-(4-Hydroxyphenyl)acrylic acid, (E)-4-Hydroxycinnamic acid, (E)-p-Coumaric acid, (E)-p-Coumaric acid, (E)-p-Hydroxycinnamic acid, Naringeninic acid, p-Hydroxy-trans-cinnamic acid, p-trans-Coumaric acid, trans-3-(4-Hydroxyphenyl)-2-propenoic acid, trans-4-Coumaric acid, trans-4-Hydroxycinnamic acid, trans-p-Coumaric acid, trans-p-Coumarinic acid, trans-p-Hydroxycinnamic acid
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\c1ccc(O)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)/b6-3+
Formula Weight 164.0473
Percent Purity >95%
Chemical Name or Material trans-p-Coumaric acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.