missing translation for 'onlineSavingsMsg'
Learn More

trans-Styrylacetic acid, 96%

Product Code. 10414155 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10414155 1 g Glass bottle 1g
10205600 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414155 Supplier Thermo Scientific Acros Supplier No. 168100010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1914-58-5 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1914-58-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002783
InChI Key PSCXFXNEYIHJST-QPJJXVBHSA-N
Synonym trans-styrylacetic acid, e-4-phenylbut-3-enoic acid, 3e-4-phenylbut-3-enoic acid, 4-phenyl-3-butenoic acid, 3-butenoic acid, 4-phenyl, 3-butenoic acid, 4-phenyl-, 3e, styrylacetic acid, e-4-phenyl-3-butenoic acid, e-styrylacetic acid
PubChem CID 5370625
SMILES OC(=O)C\C=C\C1=CC=CC=C1

Specifications

Melting Point 82°C to 88°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2CO2H
Quantity 1 g
Beilstein 09, 612
IUPAC Name (3E)-4-phenylbut-3-enoic acid
Formula Weight 162.19
Percent Purity 96%
Physical Form Crystalline Powder or Needles
Chemical Name or Material trans-Styrylacetic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • trans-Styrylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.