missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,4-Diphenyl-1,3-butadiene, 99%

Product Code. 10655342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10655342 100 g Glass bottle 100g
10570581 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10655342 Supplier Thermo Scientific Acros Supplier No. 117221000
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 538-81-8 | C16H14 | 206.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-81-8
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00004791
InChI Key JFLKFZNIIQFQBS-FNCQTZNRSA-N
Synonym bistyryl, 1,4-diphenyl-1,3-butadiene, 1,4-diphenylbutadiene, trans,trans-1,4-diphenyl-1,3-butadiene, 1,4-diphenylerythrene, distyryl, trans,trans-1,4-diphenylbuta-1,3-diene, 1,4-diphenylbuta-1,3-diene, 1,1'-1e,3e-buta-1,3-diene-1,4-diyldibenzene, 1,3-butadiene, 1,4-diphenyl
PubChem CID 641683
ChEBI CHEBI:35100
IUPAC Name [(1E,3E)-4-phenylbuta-1,3-dienyl]benzene
SMILES C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1

Specifications

Melting Point 151°C to 154°C
Color White to Yellow
Boiling Point 350°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH=CHCH=CHC6H5
Quantity 100 g
Beilstein 05, 676
Fieser 01,337
Merck Index 15, 3350
Solubility Information Solubility in water: negligible
Formula Weight 206.29
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.