missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-Dibenzylideneacetone, 98%

Product Code. 10407893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10407893 5 g Glass bottle 5g
10095740 25 g Glass bottle 25g
10371061 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407893 Supplier Thermo Scientific Acros Supplier No. 409000050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 35225-79-7 | C17H14O | 234.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35225-79-7
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.30
MDL Number MFCD00004790
InChI Key WMKGGPCROCCUDY-PHEQNACWSA-N
Synonym dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one
PubChem CID 640180
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
SMILES O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 107°C to 114°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula C6H5CH=CHC(O)CH=CHC6H5
Quantity 5 g
Beilstein 07, IV, 1747
Merck Index 15, 3010
Solubility Information Solubility in water: negliglible. Other solubilities: soluble in acetone and chloroform,slightly soluble in alcohol and ether
Formula Weight 234.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material trans, trans-Dibenzylideneacetone, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.