Learn More
Transcription factor E3 Antibody (CL12810) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18574F
Product Code. 30291076
  • 770.00 € / 0.10mL
Description
Transcription factor E3 Monoclonal antibody specifically detects Transcription factor E3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transcription factor E3
Monoclonal
FITC
BHLHE33, bHLHe33transcription factor E3, Class E basic helix-loop-helix protein 33, RCCP2, TFEA, transcription factor binding to IGHM enhancer 3, transcription factor E family, member A, Transcription factor for IgH enhancer
This antibody was developed using a recombinant protein derived from P19532, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Immunology
7030
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL12810
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified