missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transketolase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12646-50ul
This item is not returnable.
View return policy
Description
Transketolase Polyclonal antibody specifically detects Transketolase in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Transketolase | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase | |
| Produced in rabbits immunized with E. coli-derived Human Transketolase fragment | |
| 50 μg | |
| Stem Cell Markers | |
| 7086 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction