Learn More
enQuireBio™ TranslationBlocker Human Estrogen receptor siRNA
Blocks transcription and translation of Estrogen receptor from Human.
Brand:  enQuireBio™ QX14-2nmol
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16039171
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
NP_000116
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
2099
2 nmol
Human
MCF7 Human Adenocarcinoma Cells from Breast Cancer.
ESR1
siRNA
ESRA, ESR, NR3A1, ER, ESTRR, Era
Lyophilized