missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human VEGF siRNA
Blocks transcription and translation of VEGF from Human.
Brand: enQuireBio™ QX24-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_003367 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7422 | |
2 nmol | |
Human |
Pc-3 Human Prostate Adenocarcinoma Cells. | |
VEGFA | |
siRNA | |
MVCD1, VPF, VEGFA, VEGF-A | |
Lyophilized |