missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human VEGF siRNA
Blocks transcription and translation of VEGF from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_003367 |
---|---|
Cell Lines Tested | Pc-3 Human Prostate Adenocarcinoma Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | VEGFA |
Gene ID (Entrez) | 7422 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059191
|
enQuireBio™
QX24-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049191
|
enQuireBio™
QX24-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003367 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7422 | |
MVCD1, VPF, VEGFA, VEGF-A | |
Lyophilized |
Pc-3 Human Prostate Adenocarcinoma Cells. | |
VEGFA | |
siRNA | |
Human |