missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Transmissible Gastroenteritis Virus Monoclonal Antibody (TGEV11-314.6), Invitrogen™
Mouse Monoclonal Antibody
424.00 € - 1052.00 €
Specifications
| Antigen | Transmissible Gastroenteritis Virus |
|---|---|
| Clone | TGEV11-314.6 |
| Concentration | 0.65 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17800156
|
Invitrogen™
MA5-46182 |
100 μg |
424.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11593393
|
Invitrogen™
MA110784 |
500 μg |
566.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17830276
|
Invitrogen™
MA5-46147 |
1 mg |
1052.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10784 detects Transmissible Gastroenteritis Virus. MA1-10784 has been successfully used in ELISA applications. The MA1-10784 immunogen is TGEV-infected ST cells.
Transmissible gastroenteritis virus or Transmissible gastroenteritis coronavirus is a coronavirus which infects pigs. It is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the APN receptor.Specifications
| Transmissible Gastroenteritis Virus | |
| 0.65 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| TGEV-infected ST cells. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| TGEV11-314.6 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| TGV | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title