Learn More
Transthyretin/Prealbumin Antibody (CL0290) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52905JF646
Product Code. 30295371
  • 770.00 € / 0.10mL
Description
Transthyretin/Prealbumin Monoclonal antibody specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transthyretin/Prealbumin
Monoclonal
Janelia Fluor 646
ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
This antibody was developed using a recombinant protein derived from P02766, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
7276
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0290
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified