missing translation for 'onlineSavingsMsg'
Learn More

Trelagliptin, MedChemExpress

Product Code. 16492587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492587 50 mg 50mg
16466756 100 mg 100mg
16459077 5 mg 5mg
16406498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492587 Supplier Medchem Express Supplier No. HY15408
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Trelagliptin (SYR-472) is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM).

Trelagliptin (SYR-472) is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM).

Specifications

Chemical Name or Material Trelagliptin
CAS 865759-25-7
Color White
Purity Grade Notes Research
Molecular Formula C18H20FN5O2
Quantity 50 mg
Synonym SYR-472
Solubility Information DMSO : ≥ 50 mg/mL (139.91 mM)
SMILES N#CC1=CC=C(F)C=C1CN(C(N2C)=O)C(N3C[C@H](N)CCC3)=CC2=O
Molecular Weight (g/mol) 357.38
Formula Weight 357.38
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.