missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2 Antibody (2B5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-07101SS
This item is not returnable.
View return policy
Description
TREM2 Monoclonal specifically detects TREM2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TREM2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1000, Flow Cytometry 2-5ug, ELISA 1:500-1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin | |
| Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2B5 | |
| Unconjugated | |
| Q9NZC2 | |
| TREM2 | |
| Recombinant human TREM2 (19-161aa) purified from E. coli (NP_061838). | |
| 0.02 mL | |
| Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience | |
| 54209 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction