missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Trenbolone Polyclonal Antibody
GREENER_CHOICE

Product Code. 11514270
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11514270 1 mg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11514270 Supplier Invitrogen™ Supplier No. PA175009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Working Concentration: 0.25 μg/mL. Sensitivity: 10 ng/mL Alpha trenbolone produces 93% inhibition in a competitive ELISA employing 17αTrenbolone polyclonal antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Trenbolone
Applications ELISA
Classification Polyclonal
Concentration 9.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias 17-beta-Trenbolone; 17beta-Trenbolone; C18H22O2; Trenbolonum; Trienbolone
Host Species Sheep
Immunogen 17a-Trenbolone(3)-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.