missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H4K20me3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540089
This item is not returnable.
View return policy
Description
H4K20me3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA, ARRAY
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| H4K20me3 | |
| Polyclonal | |
| Unconjugated | |
| H4C1 | |
| CELE_T10C6.11; F07B7.11; F07B7.4; F45F2.12; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K20; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4c11; H4C12; H4C13; H4c14; H4C15; H4C2; H4C3; H4C4; H4C5; H4c6; H4C8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K20; H4Lys20me3; H4M; his-20; his-22; his-4; his-52; his-54; his-8; HIST1H4; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; Hist1h4h; Hist1h4i; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; Histone H2B 2; Histone H4; Histone H4A; Histone H4K20me3; histone IV, family 2; K06C4.12; K06C4.4; methyl Histone 3; methyl Histone H3; T10C6.11; Trimethyl H4; Tri-methyl H4; Trimethyl Histone H4; Tri-methyl Histone H4; Tri-Methyl-H4 K20; Tri-Methyl-H4 Lys20; Tri-Methyl-Histone H4 K20; Tri-Methyl-Histone H4 Lys20; X04652 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121504, 8294, 8359, 8363, 8366, 8367, 8370 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Peptide Array, ELISA, Western Blot, Immunocytochemistry | |
| 1.07 mg/mL | |
| PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 | |
| P62805 | |
| H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9 | |
| KLH-conjugated synthetic peptide corresponding to human the region of histone H4 containing the trimethylated lysine 20 (H4K20me3). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction