missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, MedChemExpress

Product Code. 16408496
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408496 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408496 Supplier Medchem Express Supplier No. HYB0896
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Triacetin is an artificial chemical compound, is the triester of glycerol and acetic acid, and is the second simplest fat after triformin.

Triacetin is an artificial chemical compound, is the triester of glycerol and acetic acid, and is the second simplest fat after triformin.

Specifications

Chemical Name or Material Triacetin
Physical Form Liquid
CAS 102-76-1
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H14O6
Synonym Glyceryl triacetate 1,2,3-Triacetoxypropane
Solubility Information DMSO : ≥ 2.3 mg/mL (10.54 mM)
SMILES CC(OCC(OC(C)=O)COC(C)=O)=O
Molecular Weight (g/mol) 218.2
Formula Weight 218.2
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.