missing translation for 'onlineSavingsMsg'
Learn More

Triamiphos, Dr. Ehrenstorfer

Product Code. 30320402
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320402 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320402 Supplier Dr. Ehrenstorfer Supplier No. DREC17640000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triamiphos
CAS 1031-47-6
Analyte or Component Names Triamiphos
Shipping Condition Room Temperature
Molecular Formula C12 H19 N6 O P
Certifications/Compliance ISO 17034
Synonym P-(5-Amino-3-phenyl-1H-1,2,4-triazol-1-yl)-N,N,N',N'-tetramethylphosphonic diamide, 5-Amino-1-[bis(dimethylamino)phosphinyl]-3-phenyl-1,2,4-triazole, 5-Amino-1-bis(dimethylamido)phosphoryl-3-phenyl-1,2,4-triazole, 5-Amino-3-phenyl-1,2,4-triazol-1-yl-N,N,N',N'-tetramethylphosphodiamide, NSC 232670, Triamiphos, WP 155, Wepsyn, Wepsyn 155
InChI Formula InChI=1S/C12H19N6OP/c1-16(2)20(19,17(3)4)18-12(13)14-11(15-18)10-8-6-5-7-9-10/h5-9H,1-4H3,(H2,13,14,15)
Solution Type Neat
SMILES CN(C)P(=O)(N(C)C)n1nc(nc1N)c2ccccc2
IUPAC Name 2-[bis(dimethylamino)phosphoryl]-5-phenyl-1,2,4-triazol-3-amine
Industry Type Environmental Testing
Molecular Weight (g/mol) 294.29
Quantity 25 mg
Formula Weight 294.1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.