missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triazine Pesticides Mixture 926 100 μg/mL in Acetone, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Ametryn∣Atraton∣Atrazine∣Prometon∣Prometryn∣Propazine∣Secbumeton∣Simazine∣Simetryn∣Terbuthylazine∣Terbutryn |
| CAS | 834-12-8, 1610-17-9, 1912-24-9, 1610-18-0, 7287-19-6, 139-40-2 |
| Analyte or Component Names | Ametryn∣Atraton∣Atrazine∣Prometon∣Prometryn∣Propazine∣Secbumeton∣Simazine∣Simetryn∣Terbuthylazine∣Terbutryn |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Ametryn 100 μg/mL∣Atraton 100 μg/mL∣Atrazine 100 μg/mL∣Prometon 100 μg/mL∣Prometryn 100 μg/mL∣Propazine 100 μg/mL∣Secbumeton 100 μg/mL∣Simazine 100 μg/mL∣Simetryn 100 μg/mL∣Terbuthylazine 100 μg/mL∣Terbutryn 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction