missing translation for 'onlineSavingsMsg'
Learn More

Tribenzylamine, 99+%

Product Code. 10071810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10071810 100 g 100g
10276200 250 g 250g
10239443 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071810 Supplier Thermo Scientific Acros Supplier No. 156471000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 620-40-6 | C21H21N | 287.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 620-40-6
Molecular Formula C21H21N
Molecular Weight (g/mol) 287.41
MDL Number MFCD00004773
InChI Key MXHTZQSKTCCMFG-UHFFFAOYSA-N
Synonym tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052
PubChem CID 24321
IUPAC Name N,N-dibenzyl-1-phenylmethanamine
SMILES C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1

Specifica

Melting Point 90°C to 94°C
Color White to Cream
Boiling Point 380°C to 390°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5CH2)3N
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol,soluble in toluene
Formula Weight 287.39
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Tribenzylamine, 99+%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Tribenzylamine
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.