missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(4-fluorophenyl)stannane, 95%, Thermo Scientific™

Product Code. 10305302
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10305302 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10305302 Supplier Thermo Scientific Maybridge Supplier No. SP00184DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17151-47-2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C18H31FSn
MDL Number MFCD01313817
Quantity 1 g
Synonym tributyl 4-fluorophenyl stannane, tributyl-4-fluorophenyl stannane, 4-fluoro-tri-n-butylstannyl benzene, 4-fluorophenyltributyltin, 4-fluorophenyl tributylstannane, 4-fluorophenyl-tri-n-butylstannane, 4-tri-n-butylstannyl fluorobenzene, 4-fluoro-tributylstannyl-benzene, stannane, tributyl 4-fluorophenyl, tri-n-butyl-4-fluorophenyl stannane
InChI Key FLPKMHDTSOPPOJ-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=C(C=C1)F
IUPAC Name tributyl-(4-fluorophenyl)stannane
Molecular Weight (g/mol) 385.154
PubChem CID 2737524
Formula Weight 385.15
Percent Purity 95%
Chemical Name or Material Tributyl(4-fluorophenyl)stannane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.