missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylborane, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
800mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Color | Colorless to Yellow |
| Linear Formula | [CH3(CH2)3]3B |
| Concentration | 0.94 to 1.06M |
| Fieser | 10,410 |
| Density | 0.8470g/mL |
| Formula Weight | 182.16 |
| Packaging | AcroSeal™ Glass Bottle |
| Specific Gravity | 0.847 |
| Quantity | 800 mL |
| Flash Point | −20°C |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction